Assay Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027432.3 | NP_081708.1 | 6 - 7 | 707 | 144 | |
| GenBank mRNA | AK012014.1 | - | 6 - 7 | 698 | 144 | |
| AK032897.1 | - | 4 - 5 | 246 | 144 | ||
| AK048488.1 | - | 6 - 7 | 730 | 144 | ||
| AK050391.1 | - | 6 - 7 | 707 | 144 | ||
| AK078856.1 | - | 7 - 8 | 666 | 144 | ||
| AK131608.1 | - | 5 - 6 | 618 | 144 | ||
| AK161812.1 | - | 6 - 7 | 682 | 144 | ||
| AK163839.1 | - | 702 | 144 | |||
| AK168006.1 | - | 6 - 7 | 709 | 144 | ||
| AK168752.1 | - | 6 - 7 | 729 | 144 | ||
| AK169128.1 | - | 6 - 7 | 708 | 144 | ||
| BC005755.1 | - | 6 - 7 | 662 | 2654151 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131608.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wdr77
Genomic Map