Assay Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027432.3 | NP_081708.1 | 7 - 8 | 778 | 74 | |
| GenBank mRNA | AK012014.1 | - | 7 - 8 | 769 | 74 | |
| AK032897.1 | - | 5 - 6 | 317 | 74 | ||
| AK048488.1 | - | 7 - 8 | 801 | 74 | ||
| AK050391.1 | - | 7 - 8 | 778 | 74 | ||
| AK078856.1 | - | 8 - 9 | 737 | 74 | ||
| AK131608.1 | - | 6 - 7 | 689 | 74 | ||
| AK161812.1 | - | 7 - 8 | 753 | 74 | ||
| AK163839.1 | - | 773 | 74 | |||
| AK168006.1 | - | 7 - 8 | 780 | 74 | ||
| AK168752.1 | - | 7 - 8 | 800 | 74 | ||
| AK169128.1 | - | 7 - 8 | 779 | 74 | ||
| BC005755.1 | - | 7 - 8 | 733 | 2654151 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wdr77
Genomic Map