Assay Details
| Gene Symbol: Tmigd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027025.4 | NP_081301.2 | 2 - 3 | 258 | 74 | |
| GenBank mRNA | AK005581.1 | - | 2 - 3 | 210 | 74 | |
| BC038878.1 | - | 1 - 2 | 138 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmigd3
Genomic Map