| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_001252555.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_001252555.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_001252556.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_001252556.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_010363.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
NM_010363.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006372.mm.1 |
TC1200000869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_001252555.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_001252555.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_001252556.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_001252556.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_010363.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
NM_010363.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006371.mm.1 |
TC1200000869.mm.1 |
MTA TC |