Assay Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011479.3 | NP_035609.1 | 10 - 11 | 1491 | 64 | |
| GenBank mRNA | AK044917.1 | - | 10 - 11 | 1492 | 64 | |
| AK057644.1 | - | 10 - 11 | 1527 | 64 | ||
| AK080181.1 | - | 4 - 5 | 1689 | 64 | ||
| AK089019.1 | - | 10 - 11 | 1485 | 64 | ||
| AK133728.1 | - | 4 - 5 | 552 | 64 | ||
| AK139681.1 | - | 2 - 3 | 194 | 64 | ||
| AK159346.1 | - | 10 - 11 | 1492 | 64 | ||
| AK172965.1 | - | 3 - 4 | 287 | 64 | ||
| BC003227.1 | - | 10 - 11 | 1524 | 3483158 | 64 | |
| U27455.1 | - | 10 - 11 | 1459 | 64 | ||
| X95642.1 | - | 10 - 11 | 1462 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc2
Genomic Map