Assay Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011479.3 | NP_035609.1 | 6 - 7 | 899 | 93 | |
| GenBank mRNA | AK044917.1 | - | 6 - 7 | 900 | 93 | |
| AK057644.1 | - | 6 - 7 | 935 | 93 | ||
| AK089019.1 | - | 6 - 7 | 893 | 93 | ||
| AK159346.1 | - | 6 - 7 | 900 | 93 | ||
| BC003227.1 | - | 6 - 7 | 932 | 3483158 | 93 | |
| U27455.1 | - | 6 - 7 | 867 | 93 | ||
| X95642.1 | - | 6 - 7 | 870 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc2
Genomic Map