Assay Details
| Gene Symbol: Sgsm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134091.2 | NP_598852.2 | 6 - 7 | 662 | 94 | |
| GenBank mRNA | AK170806.1 | - | 6 - 7 | 629 | 94 | |
| AY382616.1 | - | 6 - 7 | 616 | 94 | ||
| BC018197.1 | - | 6 - 7 | 622 | 4190407 | 94 | |
| BC024122.1 | - | 6 - 7 | 622 | 5097970 | 94 | |
| BC025561.1 | - | 6 - 7 | 600 | 5322980 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm3
Genomic Map