Assay Details
| Gene Symbol: Prickle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033217.4 | NP_001028389.1 | 4 - 5 | 640 | 66 | |
| GenBank mRNA | AK147883.1 | - | 4 - 5 | 653 | 66 | |
| AK153711.1 | - | 4 - 5 | 641 | 66 | ||
| BC023970.1 | - | 1 - 2 | 233 | 66 | ||
| BC117892.1 | - | 4 - 5 | 612 | 40096843 | 66 | |
| BC117893.1 | - | 4 - 5 | 612 | 40096845 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023970.1 transcripts.
Set Membership: |
|

Prickle1
Genomic Map