Assay Details
| Gene Symbol: Prickle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033217.4 | NP_001028389.1 | 5 - 6 | 843 | 134 | |
| GenBank mRNA | AK147883.1 | - | 5 - 6 | 856 | 134 | |
| AK153711.1 | - | 5 - 6 | 844 | 134 | ||
| BC117892.1 | - | 5 - 6 | 815 | 40096843 | 134 | |
| BC117893.1 | - | 5 - 6 | 815 | 40096845 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prickle1
Genomic Map