Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 18 - 19 | 6606 | 70 | |
| GenBank mRNA | BQ180311.1 | - | 3 - 4 | 342 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Celsr1
Genomic Map