Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 20 - 21 | 6940 | 65 | |
| GenBank mRNA | AF031572.1 | - | 20 - 21 | 6940 | 65 | |
| AK220170.1 | - | 19 - 20 | 2834 | 65 | ||
| BQ942572.1 | - | 1 - 2 | 124 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celsr1
Genomic Map