Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 31 - 32 | 8458 | 118 | |
| GenBank mRNA | CV561900.1 | - | 3 - 4 | 230 | 118 | |
More Information
Set Membership: |
|

Celsr1
Genomic Map