Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 33 - 34 | 8843 | 72 | |
| GenBank mRNA | AF031572.1 | - | 33 - 34 | 8843 | 72 | |
| AK220170.1 | - | 32 - 33 | 4734 | 72 | ||
| CV561900.1 | - | 5 - 6 | 615 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celsr1
Genomic Map