Assay Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145475.4 | NP_663450.3 | 3 - 4 | 873 | 94 | |
| GenBank mRNA | AB079067.1 | - | 3 - 4 | 380 | 94 | |
| AK042077.1 | - | 3 - 4 | 464 | 94 | ||
| AK052269.1 | - | 3 - 4 | 639 | 94 | ||
| AK169528.1 | - | 3 - 4 | 458 | 94 | ||
| AK188338.1 | - | 1 - 2 | 103 | 94 | ||
| BC094253.1 | - | 3 - 4 | 872 | 6400872 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cerk
Genomic Map