Assay Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB079067.1 | - | 10 - 11 | 1128 | 67 | |
| AK129416.1 | - | 7 - 8 | 763 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB079067.1 | - | 10 - 11 | 1128 | 67 | |
| AK129416.1 | - | 7 - 8 | 763 | 67 | ||
Set Membership: |
|