Assay Details
| Gene Symbol: Plp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290561.1 | NP_001277490.1 | 2 - 3 | 357 | 120 | |
| NM_001290562.1 | NP_001277491.1 | 2 - 3 | 357 | 120 | ||
| NM_011123.3 | NP_035253.1 | 2 - 3 | 357 | 120 | ||
| GenBank mRNA | AK077564.1 | - | 2 - 3 | 313 | 120 | |
| AK134554.1 | - | 2 - 3 | 348 | 120 | ||
| AK160872.1 | - | 2 - 3 | 352 | 120 | ||
| AK192772.1 | - | 2 - 3 | 240 | 120 | ||
| BC027010.1 | - | 2 - 3 | 267 | 5364736 | 120 | |
| M14674.1 | - | 1 - 2 | 191 | 120 | ||
| M15442.1 | - | 2 - 3 | 336 | 120 | ||
| M16472.1 | - | 1 - 2 | 191 | 120 | ||
| M20752.1 | - | 1 - 2 | 191 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plp1
Genomic Map