Assay Details
| Gene Symbol: Sept6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177323.2 | NP_001170794.1 | 4 - 5 | 807 | 69 | |
| NM_001177324.1 | NP_001170795.1 | 4 - 5 | 810 | 69 | ||
| NM_001253706.1 | NP_001240635.1 | 4 - 5 | 810 | 69 | ||
| NM_019942.5 | NP_064326.2 | 4 - 5 | 810 | 69 | ||
| GenBank mRNA | AB023622.1 | - | 4 - 5 | 574 | 69 | |
| AK012858.1 | - | 5 - 6 | 604 | 69 | ||
| AK048455.1 | - | 4 - 5 | 632 | 69 | ||
| AK088406.1 | - | 4 - 5 | 590 | 69 | ||
| AK088614.1 | - | 4 - 5 | 573 | 69 | ||
| AK162755.1 | - | 4 - 5 | 610 | 69 | ||
| AK172894.1 | - | 4 - 5 | 591 | 69 | ||
| BC010489.1 | - | 4 - 5 | 596 | 4168214 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept6
Genomic Map