Assay Details
| Gene Symbol: Sept6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177323.2 | NP_001170794.1 | 7 - 8 | 1244 | 67 | |
| NM_001177324.1 | NP_001170795.1 | 7 - 8 | 1247 | 67 | ||
| NM_001253706.1 | NP_001240635.1 | 7 - 8 | 1247 | 67 | ||
| NM_019942.5 | NP_064326.2 | 7 - 8 | 1247 | 67 | ||
| GenBank mRNA | AB023622.1 | - | 7 - 8 | 1011 | 67 | |
| AK012858.1 | - | 8 - 9 | 1038 | 67 | ||
| AK048455.1 | - | 7 - 8 | 1069 | 67 | ||
| AK088406.1 | - | 7 - 8 | 1027 | 67 | ||
| AK160884.1 | - | 7 - 8 | 1041 | 67 | ||
| AK162755.1 | - | 7 - 8 | 1047 | 67 | ||
| AK172894.1 | - | 7 - 8 | 1028 | 67 | ||
| BC010489.1 | - | 7 - 8 | 1033 | 4168214 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept6
Genomic Map