Assay Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026573.2 | NP_080849.1 | 1 - 2 | 244 | 66 | |
| GenBank mRNA | AK011606.1 | - | 1 - 2 | 242 | 66 | |
| AK019989.1 | - | 1 - 2 | 222 | 66 | ||
| AK145431.1 | - | 1 - 2 | 247 | 66 | ||
| AK162222.1 | - | 1 - 2 | 240 | 66 | ||
| BC058761.1 | - | 1 - 2 | 225 | 66 | ||
| BC117769.1 | - | 1 - 2 | 262 | 40091025 | 66 | |
| BC117770.1 | - | 1 - 2 | 262 | 40091027 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf3b
Genomic Map