Assay Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026573.2 | NP_080849.1 | 3 - 4 | 459 | 138 | |
| GenBank mRNA | AK011606.1 | - | 3 - 4 | 457 | 138 | |
| AK019989.1 | - | 3 - 4 | 437 | 138 | ||
| AK145431.1 | - | 3 - 4 | 462 | 138 | ||
| AK162222.1 | - | 3 - 4 | 455 | 138 | ||
| BC047049.1 | - | 1 - 2 | 118 | 138 | ||
| BC058761.1 | - | 3 - 4 | 440 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upf3b
Genomic Map