Assay Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026573.2 | NP_080849.1 | 4 - 5 | 559 | 113 | |
| GenBank mRNA | AK011606.1 | - | 4 - 5 | 557 | 113 | |
| AK019989.1 | - | 4 - 5 | 537 | 113 | ||
| AK145431.1 | - | 4 - 5 | 562 | 113 | ||
| AK162222.1 | - | 4 - 5 | 555 | 113 | ||
| BC047049.1 | - | 2 - 3 | 218 | 113 | ||
| BC058761.1 | - | 4 - 5 | 540 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upf3b
Genomic Map