Assay Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026573.2 | NP_080849.1 | 8 - 9 | 1065 | 73 | |
| GenBank mRNA | AK145431.1 | - | 8 - 9 | 1068 | 73 | |
| BC047049.1 | - | 6 - 7 | 724 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upf3b
Genomic Map