| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200014700.mm.1 |
NM_027744.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014700.mm.1 |
NM_027744.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014700.mm.1 |
TC1200002119.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014696.mm.1 |
NM_027744.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014696.mm.1 |
NM_027744.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014696.mm.1 |
TC1200002119.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014697.mm.1 |
NM_027744.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014697.mm.1 |
NM_027744.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014697.mm.1 |
TC1200002119.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200014699.mm.1 |
NM_027744.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014699.mm.1 |
NM_027744.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200014699.mm.1 |
TC1200002119.mm.1 |
MTA TC |