Assay Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019816.1 | NP_062790.1 | 3 - 4 | 489 | 78 | |
| GenBank mRNA | AF222801.1 | - | 3 - 4 | 489 | 78 | |
| AK077789.1 | - | 3 - 4 | 513 | 78 | ||
| AK141334.1 | - | 3 - 4 | 487 | 78 | ||
| BC025080.1 | - | 3 - 4 | 476 | 2648117 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aatf
Genomic Map