Assay Details
| Gene Symbol: Fndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081416.1 | NP_001074885.1 | 18 - 19 | 4958 | 96 | |
| GenBank mRNA | AK003938.1 | - | 1 - 2 | 79 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fndc1
Genomic Map