Assay Details
| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081020.1 | NP_001074489.1 | 19 - 20 | 2841 | 90 | |
| GenBank mRNA | AK030315.1 | - | 19 - 20 | 3028 | 90 | |
| AK034499.1 | - | 2 - 3 | 226 | 90 | ||
| AK044528.1 | - | 19 - 20 | 2857 | 90 | ||
| BC172116.1 | - | 19 - 20 | 2778 | 90 | ||
More Information
Set Membership: |
|

Adamts6
Genomic Map