Assay Details
| Gene Symbol: Rbm41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172147.1 | NP_001165618.1 | 4 - 5 | 492 | 102 | |
| NM_001172148.1 | NP_001165619.1 | 3 - 4 | 410 | 102 | ||
| NM_001290630.1 | NP_001277559.1 | 3 - 4 | 410 | 102 | ||
| NM_153586.2 | NP_705814.1 | 3 - 4 | 410 | 102 | ||
| GenBank mRNA | AK040213.1 | - | 3 - 4 | 350 | 102 | |
| AK077972.1 | - | 3 - 4 | 550 | 102 | ||
| AK080041.1 | - | 1 - 2 | 176 | 102 | ||
| AK084514.1 | - | 3 - 4 | 345 | 102 | ||
| AK084629.1 | - | 3 - 4 | 345 | 102 | ||
| AK141075.1 | - | 3 - 4 | 345 | 102 | ||
| AK163147.1 | - | 2 - 3 | 329 | 102 | ||
| AK164496.1 | - | 4 - 5 | 440 | 102 | ||
| AK166005.1 | - | 3 - 4 | 345 | 102 | ||
| AK166229.1 | - | 358 | 102 | |||
| BC037024.1 | - | 3 - 4 | 377 | 5368185 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm41
Genomic Map