Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172449.2 | NP_766037.2 | 2 - 3 | 1291 | 84 | |
| GenBank mRNA | AK043747.1 | - | 2 - 3 | 1293 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bzrap1
Genomic Map