Assay Details
| Gene Symbol: Atf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287180.1 | NP_001274109.1 | 2 - 3 | 224 | 101 | |
| NM_009716.3 | NP_033846.2 | 1 - 2 | 507 | 101 | ||
| GenBank mRNA | AA003141.1 | - | 2 - 3 | 223 | 101 | |
| AK138657.1 | - | 1 - 2 | 192 | 101 | ||
| AK144777.1 | - | 1 - 2 | 193 | 101 | ||
| AK146193.1 | - | 1 - 2 | 195 | 101 | ||
| AK150231.1 | - | 1 - 2 | 194 | 101 | ||
| AK155254.1 | - | 1 - 2 | 193 | 101 | ||
| AK156298.1 | - | 1 - 2 | 187 | 101 | ||
| AK157839.1 | - | 1 - 2 | 162 | 101 | ||
| BC085169.1 | - | 1 - 2 | 398 | 30551592 | 101 | |
| M94087.1 | - | 1 - 2 | 174 | 101 | ||
| X61507.1 | - | 1 - 2 | 185 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138657.1, AK144777.1, AK146193.1, AK155254.1, AK156298.1, AK157839.1, BC085169.1, M94087.1, NM_001287180.1, NM_009716.3, X61507.1 transcripts.
Set Membership: |
|

Atf4
Genomic Map