Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080769.1 | NP_001074238.1 | 16 - 17 | 3791 | 78 | |
| GenBank mRNA | AK089790.1 | - | 16 - 17 | 3819 | 78 | |
| AK144200.1 | - | 11 - 12 | 3087 | 78 | ||
| AK220293.1 | - | 16 - 17 | 3791 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uhrf1bp1
Genomic Map