Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080769.1 | NP_001074238.1 | 8 - 9 | 1269 | 93 | |
| GenBank mRNA | AK220293.1 | - | 8 - 9 | 1269 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uhrf1bp1
Genomic Map