Assay Details
| Gene Symbol: Slc8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167920.1 | NP_001161392.1 | 3 - 4 | 2491 | 63 | |
| GenBank mRNA | AK139260.1 | - | 3 - 4 | 2478 | 63 | |
| AK165197.1 | - | 3 - 4 | 2277 | 63 | ||
| BC052435.1 | - | 3 - 4 | 2262 | 6837128 | 63 | |
| BC080862.1 | - | 3 - 4 | 1979 | 30357598 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a3
Genomic Map