Assay Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113386.1 | NP_001106857.1 | 4 - 5 | 660 | 98 | |
| NM_013584.2 | NP_038612.1 | 3 - 4 | 309 | 98 | ||
| GenBank mRNA | AK155524.1 | - | 3 - 4 | 536 | 98 | |
| BC031929.1 | - | 3 - 4 | 302 | 4159053 | 98 | |
| BY716975.1 | - | 3 - 4 | 527 | 98 | ||
| D17444.1 | - | 3 - 4 | 316 | 98 | ||
| D26177.1 | - | 3 - 4 | 316 | 98 | ||
| S73495.1 | - | 3 - 4 | 316 | 98 | ||
| S73496.1 | - | 3 - 4 | 316 | 98 | ||
More Information
Set Membership: |
|

Lifr
Genomic Map