Assay Details
| Gene Symbol: Ttc23l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029430.1 | NP_083706.1 | 10 - 11 | 1307 | 144 | |
| GenBank mRNA | AK015027.1 | - | 10 - 11 | 1279 | 144 | |
| BC132384.1 | - | 10 - 11 | 1307 | 144 | ||
| BC145852.1 | - | 10 - 11 | 1307 | 40131155 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969511 | BC145852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969511 | NM_029430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969511 | AK015027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969511 | BC132384.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801469 | DQ005956.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801469 | NM_001042503.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801469 | XM_001476361.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814158 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82670 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148151 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120308 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969511 | BC145852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969511 | NM_029430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969511 | AK015027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969511 | BC132384.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08282706 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120308 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08282671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08282784 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08282655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02972591 | - | - |
| Whole Mouse Genome G4122A | A_51_P143744 | BC145852.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143744 | NM_029430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143744 | AK015027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143744 | BC132384.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120308 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12270533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12104853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11996380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11860106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12285752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12086987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12423052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12423055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11838490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12247927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12171649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11996377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12143211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11838488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12171650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11996371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30010965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30011117 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10427735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427731 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431902_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10427729 | - | - |
| Mouse Genome 430 2.0 Array | 1431902_at | - | - |
| Mouse Genome 430 2.0 Array | 1431902_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500009709.mm.1 | NM_029430.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009709.mm.1 | NM_029430.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009709.mm.1 | TC1500001204.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500009708.mm.1 | NM_029430.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009708.mm.1 | NM_029430.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009708.mm.1 | TC1500001204.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431902_at | - | - |
| Mouse Expression Set 430 | 1431902_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431902_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431902_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 742736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747951 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2747951 | - | - |

Ttc23l
Genomic Map