Assay Details
| Gene Symbol: Ttll8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172818.3 | NP_766406.2 | 2 - 3 | 652 | 129 | |
| GenBank mRNA | AK029745.1 | - | 2 - 3 | 653 | 129 | |
| AK030151.1 | - | 2 - 3 | 645 | 129 | ||
| AM690752.1 | - | 2 - 3 | 309 | 129 | ||
| BC116293.1 | - | 2 - 3 | 293 | 40094952 | 129 | |
| BC116294.1 | - | 2 - 3 | 293 | 40094955 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029745.1, BC116294.1 transcripts.
Set Membership: |
|

Ttll8
Genomic Map