Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 11 - 12 | 970 | 99 | |
| GenBank mRNA | AK035946.1 | - | 11 - 12 | 918 | 99 | |
| AK042787.1 | - | 11 - 12 | 913 | 99 | ||
| AK048502.1 | - | 11 - 12 | 926 | 99 | ||
| AK050932.1 | - | 11 - 12 | 853 | 99 | ||
| AK129451.1 | - | 11 - 12 | 864 | 99 | ||
| AK140709.1 | - | 11 - 12 | 971 | 99 | ||
| AK157865.1 | - | 11 - 12 | 921 | 99 | ||
| BC057139.1 | - | 6 - 7 | 438 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map