Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 14 - 15 | 1209 | 144 | |
| GenBank mRNA | AK035946.1 | - | 14 - 15 | 1157 | 144 | |
| AK042787.1 | - | 14 - 15 | 1152 | 144 | ||
| AK129451.1 | - | 14 - 15 | 1103 | 144 | ||
| AK140709.1 | - | 14 - 15 | 1210 | 144 | ||
| BC057139.1 | - | 9 - 10 | 677 | 144 | ||
More Information
Set Membership: |
|

Tmx3
Genomic Map