Assay Details
| Gene Symbol: Tmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198295.2 | NP_938037.2 | 8 - 9 | 743 | 128 | |
| GenBank mRNA | AK035946.1 | - | 8 - 9 | 691 | 128 | |
| AK042787.1 | - | 8 - 9 | 686 | 128 | ||
| AK048502.1 | - | 8 - 9 | 699 | 128 | ||
| AK050932.1 | - | 8 - 9 | 626 | 128 | ||
| AK129451.1 | - | 8 - 9 | 637 | 128 | ||
| AK140709.1 | - | 8 - 9 | 744 | 128 | ||
| AK157865.1 | - | 8 - 9 | 694 | 128 | ||
| BC057139.1 | - | 3 - 4 | 211 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmx3
Genomic Map