Assay Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144838.1 | NP_659087.1 | 6 - 7 | 682 | 93 | |
| GenBank mRNA | AK049820.1 | - | 6 - 7 | 720 | 93 | |
| AK082080.1 | - | 6 - 7 | 572 | 93 | ||
| BC017611.1 | - | 6 - 7 | 682 | 4527683 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgtb
Genomic Map