Assay Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144838.1 | NP_659087.1 | 8 - 9 | 888 | 77 | |
| GenBank mRNA | AK049820.1 | - | 8 - 9 | 926 | 77 | |
| AK082080.1 | - | 8 - 9 | 778 | 77 | ||
| BC017611.1 | - | 8 - 9 | 888 | 4527683 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgtb
Genomic Map