Assay Details
| Gene Symbol: Dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176835.2 | NP_789805.1 | 1 - 2 | 117 | 92 | |
| GenBank mRNA | AK013323.1 | - | 1 - 2 | 141 | 92 | |
| AK050294.1 | - | 1 - 2 | 95 | 92 | ||
| BC039633.1 | - | 1 - 2 | 117 | 5136672 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC039633.1, NM_176835.2 transcripts.
Set Membership: |
|

Dnajc22
Genomic Map