Assay Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024262.1 | NP_077224.1 | 1 - 2 | 1785 | 97 | |
| GenBank mRNA | AK004714.1 | - | 1 - 2 | 1778 | 97 | |
| AK031106.1 | - | 1 - 2 | 1759 | 97 | ||
| AK035089.1 | - | 1 - 2 | 752 | 97 | ||
| BC002247.1 | - | 1 - 2 | 390 | 97 | ||
| BC020005.1 | - | 1 - 2 | 1785 | 3981459 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smg8
Genomic Map