Assay Details
| Gene Symbol: Hagh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159626.1 | NP_001153098.1 | 8 - 9 | 845 | 61 | |
| NM_024284.2 | NP_077246.2 | 8 - 9 | 935 | 61 | ||
| GenBank mRNA | AK147022.1 | - | 8 - 9 | 896 | 61 | |
| AK150557.1 | - | 8 - 9 | 853 | 61 | ||
| AK151231.1 | - | 8 - 9 | 853 | 61 | ||
| AK152742.1 | - | 8 - 9 | 853 | 61 | ||
| AK165777.1 | - | 8 - 9 | 839 | 61 | ||
| BC004749.1 | - | 8 - 9 | 878 | 3583919 | 61 | |
| BC019817.1 | - | 8 - 9 | 828 | 5123648 | 61 | |
| BQ128470.1 | - | 1 - 2 | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099856 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154536 | BC019817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154536 | BQ128470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154536 | BC004749.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166097 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159111 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164152 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166657 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154536 | BC019817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154536 | BQ128470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154536 | BC004749.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127469 | BC019817.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127469 | BQ128470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127469 | AK150557.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127469 | AK151231.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127469 | AK152742.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099860 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159111 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166657 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164152 | - | - |
| Whole Mouse Genome G4122A | A_51_P127469 | BC019817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127469 | BQ128470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127469 | AK150557.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127469 | AK151231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127469 | AK152742.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108284 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110114 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164152 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159111 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13177902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13465396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13444004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13177892 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13444003 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13444006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13465419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13465411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13465406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13427806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13109024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13427799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13349311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13059736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13262039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13177913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13093695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099866 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442623 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424171_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424172_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442616 | - | - |
| Mouse Genome 430 2.0 Array | 1424171_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424171_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437519_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1424172_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424172_at | - | - |
| Mouse Genome 430 2.0 Array | 1437519_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14651_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14651_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002798.mm.1 | NM_001159626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002798.mm.1 | NM_001159626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002798.mm.1 | NM_024284.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002798.mm.1 | NM_024284.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002798.mm.1 | TC1700000354.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002796.mm.1 | NM_001159626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002796.mm.1 | NM_001159626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002796.mm.1 | NM_024284.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002796.mm.1 | NM_024284.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002796.mm.1 | TC1700000354.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002797.mm.1 | NM_001159626.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002797.mm.1 | NM_001159626.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002797.mm.1 | NM_024284.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002797.mm.1 | NM_024284.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002797.mm.1 | TC1700000354.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437519_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437519_x_at | - | - |
| Murine Genome U74v2 Set | 100042_PM_at | - | - |
| Murine Genome U74v2 Set | 100042_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437519_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437519_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424171_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424171_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424172_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424172_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 540358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 540358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000540358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734661 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734661 | - | - |
| mouse-6 v1.1 (Illumina) | scl014651.8_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 540358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014651.8_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734661 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734661 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014651.8_3-S | - | - |

Hagh
Genomic Map