Assay Details
| Gene Symbol: 1600002H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028056.1 | NP_082332.1 | 2 - 3 | 173 | 83 | |
| GenBank mRNA | AK005395.1 | - | 2 - 3 | 255 | 83 | |
| AK146539.1 | - | 2 - 3 | 167 | 83 | ||
| BC072612.1 | - | 2 - 3 | 173 | 6414405 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005395.1 transcripts.
Set Membership: |
|

1600002H07Rik
Genomic Map