| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700014302.mm.1 |
NM_028056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014302.mm.1 |
NM_028056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014302.mm.1 |
TC1700001621.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014304.mm.1 |
NM_028056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014304.mm.1 |
NM_028056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014304.mm.1 |
TC1700001621.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014309.mm.1 |
NM_028056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014309.mm.1 |
NM_028056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014309.mm.1 |
TC1700001621.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014301.mm.1 |
NM_028056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014301.mm.1 |
NM_028056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014301.mm.1 |
TC1700001621.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014307.mm.1 |
NM_028056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014307.mm.1 |
NM_028056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014307.mm.1 |
TC1700001621.mm.1 |
MTA TC |