Assay Details
| Gene Symbol: 1600002H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028056.1 | NP_082332.1 | 9 - 10 | 1197 | 65 | |
| GenBank mRNA | AK005395.1 | - | 8 - 9 | 1397 | 65 | |
| AK146539.1 | - | 9 - 10 | 1191 | 65 | ||
| BC063767.1 | - | 8 - 9 | 1115 | 65 | ||
| BC072612.1 | - | 9 - 10 | 1197 | 6414405 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128952 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137527 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31096143 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08738797 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137527 | - | - |
| Whole Mouse Genome G4122A | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | BC072612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | NM_028056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | AK005395.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193236 | AK146539.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137527 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08738797 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13093048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13093034 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13093052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13093053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13093044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31096124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13448175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13490901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13561645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13561650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31096129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13448186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448496 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448501 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428832_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448495 | - | - |
| Mouse Genome 430 2.0 Array | 1428832_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428041_at | - | - |
| Mouse Genome 430 2.0 Array | 1428832_at | - | - |
| Mouse Genome 430 2.0 Array | 1428041_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72016_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72016_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014292.mm.1 | NM_028056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014292.mm.1 | NM_028056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014292.mm.1 | TC1700001621.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014293.mm.1 | NM_028056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014293.mm.1 | NM_028056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014293.mm.1 | TC1700001621.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014294.mm.1 | NM_028056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014294.mm.1 | NM_028056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014294.mm.1 | TC1700001621.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428832_at | - | - |
| Mouse Expression Set 430 | 1428832_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428832_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428832_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 472379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2650578 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2650578 | - | - |

1600002H07Rik
Genomic Map