Assay Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144849.1 | NP_659098.1 | 5 - 6 | 659 | 58 | |
| GenBank mRNA | AK050440.1 | - | 5 - 6 | 686 | 58 | |
| AK133785.1 | - | 5 - 6 | 683 | 58 | ||
| AK143625.1 | - | 5 - 6 | 686 | 58 | ||
| BC014743.1 | - | 5 - 6 | 659 | 4503319 | 58 | |
| DQ884878.1 | - | 5 - 6 | 687 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd54
Genomic Map