Assay Details
| Gene Symbol: Gcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161712.1 | NP_001155184.1 | 6 - 7 | 861 | 107 | |
| NM_013847.4 | NP_038875.3 | 6 - 7 | 861 | 107 | ||
| GenBank mRNA | AF093403.1 | - | 6 - 7 | 819 | 107 | |
| AK007395.1 | - | 6 - 7 | 794 | 107 | ||
| AK075782.1 | - | 6 - 7 | 801 | 107 | ||
| AK076469.1 | - | 6 - 7 | 834 | 107 | ||
| AK087433.1 | - | 6 - 7 | 820 | 107 | ||
| BC024107.1 | - | 6 - 7 | 861 | 5125361 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcat
Genomic Map