Assay Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021713.2 | NP_068359.1 | 6 - 7 | 974 | 70 | |
| GenBank mRNA | AF289484.1 | - | 6 - 7 | 966 | 70 | |
| AK002656.1 | - | 6 - 7 | 949 | 70 | ||
| AK004104.1 | - | 6 - 7 | 949 | 70 | ||
| AK013232.1 | - | 6 - 7 | 920 | 70 | ||
| BC023124.1 | - | 5 - 6 | 1101 | 70 | ||
| BC028501.1 | - | 6 - 7 | 959 | 5256915 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497293 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142673 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165220 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167919 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497297 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03118314 | - | - |
| Whole Mouse Genome G4122A | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | BC028501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK002656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK004104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | AK013232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P278540 | BC023124.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111210 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12212960 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12212969 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12212968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30497293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12168522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12212970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12022467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12065260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12212968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497329 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10427209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427211 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424088_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430729_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10427205 | - | - |
| Mouse Genome 430 2.0 Array | 1430729_at | - | - |
| Mouse Genome 430 2.0 Array | 1424088_at | - | - |
| Mouse Genome 430 2.0 Array | 1424088_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430729_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60315_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60315_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008800.mm.1 | NM_021713.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008800.mm.1 | NM_021713.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008800.mm.1 | TC1500002338.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008802.mm.1 | NM_021713.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008802.mm.1 | NM_021713.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008802.mm.1 | TC1500002338.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008798.mm.1 | NM_021713.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008798.mm.1 | NM_021713.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008798.mm.1 | TC1500002338.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008799.mm.1 | NM_021713.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008799.mm.1 | NM_021713.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008799.mm.1 | TC1500002338.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424088_at | - | - |
| Mouse Expression Set 430 | 1424088_PM_at | - | - |
| Murine Genome U74v2 Set | 95651_at | - | - |
| Murine Genome U74v2 Set | 95651_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424088_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424088_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424088_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424088_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430729_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430729_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219703 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225033 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225033 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219703 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002486.1_42-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002566.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120324 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002566.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002486.1_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270113 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219703 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594821 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225033 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225033 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594821 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219703 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002566.1_2-S | - | - |

Myg1
Genomic Map