Assay Details
| Gene Symbol: Lmbr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029098.3 | NP_083374.1 | 9 - 10 | 944 | 81 | |
| GenBank mRNA | AK003656.1 | - | 9 - 10 | 876 | 81 | |
| AK030589.1 | - | 9 - 10 | 910 | 81 | ||
| AK049110.1 | - | 9 - 10 | 945 | 81 | ||
| AK143891.1 | - | 9 - 10 | 905 | 81 | ||
| AK154968.1 | - | 9 - 10 | 1078 | 81 | ||
| AK173121.1 | - | 9 - 10 | 1325 | 81 | ||
| AY052398.1 | - | 9 - 10 | 787 | 81 | ||
| BC023397.1 | - | 9 - 10 | 881 | 5043877 | 81 | |
More Information
Set Membership: |
|

Lmbr1l
Genomic Map